Learn More
Bromocyclohexane, 98%
Description
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & Light Sensitive. Store away from strong bases.
 
            Specifications
Specifications
| Melting Point | -57°C | 
| Density | 1.335 | 
| Boiling Point | 166°C to 167°C | 
| Flash Point | 62°C (143°F) | 
| Refractive Index | 1.495 | 
| Quantity | 100 g | 
| Beilstein | 1098373 | 
| Sensitivity | Light sensitive | 
| Merck Index | 14,2730 | 
| Solubility Information | Soluble in alcohol. Insoluble in water. | 
| Show More | 
Safety and Handling
| Hazard Category | H227 | 
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. | 
| Precautionary Statement | P210-P280-P370+P378q-P501c | 
| EINECSNumber | 203-622-2 | 
| RTECSNumber | GU7171700 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                