Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody

Catalog No. MS1848R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1848R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1848R1 Supplier Epredia™ Supplier No. MS1848R1

Detect S-phase cells with Epredia™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: Bu20a

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: BrdU conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Bromodeoxyuridine (BrdU) Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Bu20a
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen BrdU conjugated to BSA
Quantity 1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species All species
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.