Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodeoxyuridine/BrdU Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC7225U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7225U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7225U100 Supplier R&D Systems Supplier No. IC7225U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Bromodeoxyuridine/BrdU
Applications Flow Cytometry
Classification Monoclonal
Clone BU-1
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias 5-bromo-2-deoxyuridine, BrdU, Bromodeoxyuridine, BUdr
Host Species Mouse
Immunogen 5-iodourindine (5-IO) coupled to ovalbumin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects Bromodeoxyuridine/BrdU.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.