Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromodeoxyuridine/BrdU Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC7225S100UG
This item is not returnable.
View return policy
Description
Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Bromodeoxyuridine/BrdU | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
5-bromo-2-deoxyuridine, BrdU, Bromodeoxyuridine, BUdr | |
5-iodourindine (5-IO) coupled to ovalbumin | |
100 μg | |
Primary | |
Multi-species | |
IgG2a |
Flow Cytometry | |
BU-1 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects Bromodeoxyuridine/BrdU. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction