Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (85-2C8), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP292988MFV610
Description
Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Non-species specific samples. It is validated for Flow CytometrySpecifications
Bromodeoxyuridine/BrdU | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Non-species specific | |
Purified |
Flow Cytometry | |
85-2C8 | |
Flow Cytometry | |
Bromodeoxyuridine, BUdr | |
Bromodeoxyuridine/BrdU conjugated to hemocyanine (isolated from Helix pomatia) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction