Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (BRD/1539R), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308591JF646
Description
Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Nonspecies specific samples. It is validated for ELISASpecifications
Bromodeoxyuridine/BrdU | |
Monoclonal | |
Janelia Fluor 646 | |
Bromodeoxyuridine, BUdr | |
Bromodeoxyuridine/BrdU conjugated to KLH | |
100 μg | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Non-species specific | |
Purified |
ELISA | |
BRD/1539R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction