Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (BRD.3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452210.02MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in All Species samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Bromodeoxyuridine/BrdU | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Bromodeoxyuridine, BUdr | |
Bromodeoxyuridine (BrdU) conjugated to BSA | |
0.02 mg | |
Primary | |
Mouse, All species | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
BRD.3 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
It reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors. | |
Store at 4C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction