Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodimethylborane, 97%

Catalog No. AA3355214
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
AA3355214 25 g
1 options

Catalog No. AA3355214

Supplier: Thermo Scientific Chemicals 03355214

Only null left
Add to Cart

CAS: 5158-50-9 | C2H6BBr | 120.784 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5158-50-9
Molecular Formula C2H6BBr
Molecular Weight (g/mol) 120.784
MDL Number MFCD00000053
InChI Key ABQPEYRVNHDPIO-UHFFFAOYSA-N
Synonym dimethylboron bromide, dimethylboronbromide, borane, bromodimethyl, unii-eb60o8m22h, dmbbr, dimethylbromoborane, me2bbr, bromo dimethyl borane, acmc-20al64, ch3 2bbr
PubChem CID 123224
IUPAC Name bromo(dimethyl)borane
SMILES B(C)(C)Br

Specifications

Melting Point -129°C
Density 1.238 g/mL
Boiling Point 31°C to 32°C
Flash Point -37°C (-35°F)
Odor Acrid
Refractive Index 1.399
Quantity 25 g
UN Number UN2845
Beilstein 1730828
Sensitivity Air and moisture sensitive
Formula Weight 120.79
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bromodimethylborane
Show More Show Less
Hazard Category H224-H250-H314-H335
Hazard Statement GHS H Statement
H250-H314-H318
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.