Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Bromophenetole, 98%, Thermo Scientific™

Catalog No. p-76204
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 589-10-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00000234
InChI Key JJFOBACUIRKUPN-UHFFFAOYSA-N
Synonym 2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene
PubChem CID 68526
IUPAC Name 2-bromoethoxybenzene
SMILES C1=CC=C(C=C1)OCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.5°C to 34.0°C
Color Colorless
Boiling Point 144.0°C (40.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5550 to 1.5570
Linear Formula C6H5OCH2CH2Br
Quantity 5 g
Beilstein 06, 142
Formula Weight 201.06
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material β-Bromophenetole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-634-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.