Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue (0.1% in ca. 50% Ethanol) [for Titration], TCI America™
SDP

Catalog No. B0118500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0118500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0118500ML Supplier TCI America Supplier No. B0118500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym bromophenol blue, bromphenol blue, albutest, tetrabromphenol blue, 3,3',5,5'-tetrabromophenolsulfonphthalein, 3',3,5',5-tetrabromophenolsulfonephthalein, ccris 5487, bromophenol blue bpb, 3',3,5',5-tetrabromophenolsulfophthalein, bromphenol blue sultone form
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Quantity 500 mL
UN Number 1170
Formula Weight 669.96
Physical Form Liquid
Chemical Name or Material Bromophenol Blue (0.1% in ca. 50% Ethanol) [for Titration]
EINECSNumber (5)-3566
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.