Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromophenol Blue, 1% w/v, Reagents
Click to view available options
Quantity:
500 mL
4 L
Specifications
Specifications
Chemical Name or Material | Bromophenol Blue, 1% w/v |
Boiling Point | 100°C |
CAS | 7732-18-5, 62625-28-9 |
Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
Percent Purity | 1% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.001 |
Color | Dark Reddish Purple |
Quantity | 4 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction