Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, sodium salt, Acros Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle

Indicator

Indicator
CAS 62625-28-9
Molecular Formula C19H9Br4NaO5S
MDL Number MFCD00013793
Beilstein 19, I, 649
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 691.942
PubChem CID 3791690
Formula Weight 691.94
Grade Pure
Packaging Glass bottle
Melting Point >300.0°C
Quantity 5g
Chemical Name or Material Bromophenol Blue, sodium salt, Pure
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning