Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS

Catalog No. p-7049987
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 34725-61-6

Bromophenol Blue sodium salt is used as a tracking dye for protein and nucleic acid electrophoresis in polyacrylamide gels. It also used in the preparation of protein samples for western blotting analysis. It finds application in staining histones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
MDL Number MFCD00013793

Specifications

Melting Point >300°C
Merck Index 14,1444
Solubility Information Soluble in water.
Formula Weight 691.96
Grade ACS
Quantity 5 g
Physical Form Crystalline
Chemical Name or Material Bromophenol Blue sodium salt
EINECSNumber 252-170-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.