Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Bromostyrene, 97%, mixture of cis/trans isomers

Catalog No. p-3777702
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle

CAS: 103-64-0 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-64-0
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000185
InChI Key YMOONIIMQBGTDU-VOTSOKGWSA-N
Synonym 2-bromovinyl benzene, beta-bromostyrene, styryl bromide, 2-bromoethenyl benzene, bromostyrolene, 1-bromo-2-phenylethene, bromostyrol, bromstyrole, hyacinth base, 1-bromo-2-phenylethylene
PubChem CID 5314126
IUPAC Name [(E)-2-bromoethenyl]benzene
SMILES C1=CC=C(C=C1)C=CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 7.0°C
Color Yellow to Brown
Density 1.4200g/mL
Boiling Point 110.0°C to 112.0°C (20.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (sum of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.6060 to 1.6090
Linear Formula C6H5CH=CHBr
Quantity 5 mL
Beilstein 05, 477
Specific Gravity 1.42
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 183.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material β-Bromostyrene, Mixture of cis and trans
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 203-131-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.