Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromosuccinic acid, 96%

Catalog No. aab2282906
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 923-06-8 | C4H5BrO4 | 196.98 g/mol

Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

Solubility
Insoluble in ether.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923-06-8
Molecular Formula C4H5BrO4
Molecular Weight (g/mol) 196.98
MDL Number MFCD00004213
InChI Key QQWGVQWAEANRTK-UHFFFAOYNA-N
Synonym bromosuccinic acid, 2-bromosuccinic acid, monobromosuccinic acid, butanedioic acid, bromo, +--bromosuccinic acid, dl-bromosuccinic acid, rs-bromosuccinic acid, succinic acid, bromo, bromobutanedioic acid
PubChem CID 73557
ChEBI CHEBI:73712
IUPAC Name 2-bromobutanedioic acid
SMILES OC(=O)CC(Br)C(O)=O

Specifications

Melting Point 164°C to 166°C (decomposition)
Quantity 5 g
Beilstein 1723556
Merck Index 14,1437
Solubility Information Insoluble in ether.
Formula Weight 196.99
Percent Purity 96%
Chemical Name or Material Bromosuccinic acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-087-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.