Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoterephthalic Acid 98.0+%, TCI America™
SDP

Catalog No. B132125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B132125G 25 g
B13215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B132125G Supplier TCI America Supplier No. B132125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 586-35-6
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002403
InChI Key QPBGNSFASPVGTP-UHFFFAOYSA-N
Synonym bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9
PubChem CID 68513
IUPAC Name 2-bromoterephthalic acid
SMILES C1=CC(=C(C=C1C(=O)O)Br)C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 245.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bromoterephthalic Acid
EINECSNumber (3)-0062
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.