Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bromothymol Blue, ACS

Catalog No. p-7049986
Click to view available options
Quantity:
5 g
25 g

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol

Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Melting Point 200°C to 202°C
Beilstein 373934
Merck Index 14,1445
Solubility Information Soluble in alcohol and aqueous solutions of alkali; sparingly soluble in waterSoluble in ethanol,ether and alkalis; Slightly soluble in water.
Formula Weight 624.39
Grade ACS
Color Brown to Red
Quantity 5 g
Physical Form Crystalline
Chemical Name or Material Bromothymol Blue
EINECSNumber 200-971-2
RTECSNumber SJ450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.