Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. 01336771
Click to view available options
Quantity:
1 g
5 g
10 g
25 g

Bromothymol Blue, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromothymol Blue, ACS Reagent (Sultone) not water soluble
CAS 76-59-5
Molecular Formula C27H28Br2O5S
Synonym Dibromothymolsulfophthalein, Bromthymol Blue
Molecular Weight (g/mol) 624.3812
Grade ACS
Packaging Glass Bottle
Specific Gravity 1
Color White
Quantity 5 g
Physical Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.