Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Bromothymol Blue Solution, 0.04%
Shop All Fisherbrand Products

Click to view available options
Quantity:
10 mL
30 mL
100 mL
500 mL
1 L
Packaging:
0.04%
Lab Grade - Solution
Description
CO2/pH indicator
This product is for educational use - no C of A is available.
Specifications
Specifications
| Chemical Name or Material | Bromothymol Blue Solution, 0.04% |
| CAS | 62625-28-9, 7732-18-5 |
| Assay Percent Range | 0.04% w/v |
| Formula Weight | 646.37 |
| Packaging | Lab Grade - Solution |
| Density | 1g/mL |
| pH Color Changes | pH 6.0-7.6 (yellow-blue) |
| Color | Green |
| Quantity | 30 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction