Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue TS, 0.1% (w/v) in 50% (v/v) Alcohol, Ricca Chemical
SDP

Catalog No. 13784
missing translation for 'orderingAttributeHoverText'
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 76-59-5, 64-17-5, 67-56-1, 7732-18-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) Mixture
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
PubChem CID 6450
ChEBI CHEBI:86155

Specifications

Concentration 0.1% (w/v)
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix 50% (v/v) Alcohol
Color Orange/Red
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Bromothymol Blue, Test Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.