Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bronze gauze, alloy 220, 150 mesh woven from 0.066mm (0.0026in) in dia wire, Thermo Scientific™

Catalog No. p-7059692
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bronze gauze, Alloy 220
Name Note 150 mesh woven from 0.066mm (0.0026 in.) dia. wire
Quantity 150 x 150 mm
Mesh Size 150 Mesh
Odor Odorless
MDL Number MFCD00198187
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.