Learn More
Bronze powder, -100 mesh, 99.0% (metals basis)
Description
Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Bronze |
| Appearance | Bronze colored powder |
| Quantity | 250 g |
| Mesh Size | -100 Mesh |
| Odor | Odorless |
| Form | Powder |
| Assay Percent Range | 99% (Metals basis) |
| MDL Number | MFCD00198187 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.