Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRP44 Antibody (JCM-1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280086
Description
BRP44 Monoclonal specifically detects BRP44 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BRP44 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Immunoprecipitation | |
brain protein 44, DKFZP564B167, MGC125752, MGC125753 | |
Mouse | |
Protein G purified | |
RUO | |
25874 | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
JCM-1 | |
Unconjugated | |
PBS containing 25% glycerol | |
MPC2 | |
BRP44 peptide coupled to KLH. | |
100 μg | |
Primary | |
Recognizes endogenous human and mouse BRP44. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction