Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRP44 Antibody (JCM-1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280086R
This item is not returnable.
View return policy
Description
BRP44 Monoclonal antibody specifically detects BRP44 in Human, Mouse samples. It is validated for Western BlotSpecifications
BRP44 | |
Monoclonal | |
DyLight 550 | |
brain protein 44, DKFZP564B167, MGC125752, MGC125753 | |
BRP44 peptide coupled to KLH. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
JCM-1 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
25874 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction