Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRP44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112370
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112370 Supplier Invitrogen™ Supplier No. PA5112370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRP44 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Mediates the uptake of pyruvate into mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRP44
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MPC2
Gene Accession No. O95563, Q9D023
Gene Alias 0610006C01Rik; 2010002I07Rik; 2610205H19Rik; AA108335; brain protein 44; BRP44; ESTM43; Mitochondrial pyruvate carrier 2; Mpc2; Protein 0-44; RGD1563422
Gene Symbols MPC2
Host Species Rabbit
Immunogen Recombinant Human Mitochondrial pyruvate carrier 2 protein (1-40AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25874, 70456
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.