Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRP44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116939
Description
Positive Samples: Mouse kidney Immunogen sequence: MSAAGARGLR ATYHRLLDKV ELMLPEKLRP LYNHPAGPRT VFFWAPIMKW GLVCAGLADM.
Mediates the uptake of pyruvate into mitochondria.
Specifications
| BRP44 | |
| Polyclonal | |
| Unconjugated | |
| MPC2 | |
| 0610006C01Rik; 2010002I07Rik; 2610205H19Rik; AA108335; brain protein 44; BRP44; ESTM43; Mitochondrial pyruvate carrier 2; Mpc2; Protein 0-44; RGD1563422 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100359982, 70456 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P38718, Q9D023 | |
| MPC2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-60 of human MPC2 (NP_0562301). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction