Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRP44L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988556
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8556 Supplier Bioss Supplier No. BS8428R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRP44L (brain protein 44-like protein), also known as HSPC040 or CGI-129, is a 109 amino acid mitochondrial protein belonging to the UPF0041 family. The gene that encodes BRP44L maps to human chromosome 6. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson′s disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.

Specifications

Antigen BRP44L
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MPC1
Gene Accession No. Q9Y5U8
Gene Alias 0610006G08Rik; 3830411I18Rik; Apoptosis-regulating basic protein; Arbp; Brain protein 44-like protein; Brp44l; CGI-129; dJ68L15.3; HSPC040; HSPC040 protein; Mitochondrial pyruvate carrier 1; MPC1; MPYCD; PNAS-115
Gene Symbols MPC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BRP44L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51660
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.