Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRPF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551909 Supplier Invitrogen™ Supplier No. PA551909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLAGDEATHH TEDAAEEERL VLLENQKHLP VEEQLKLLLE RLDEVNASKQ SVGRSRRAKM IKKEMTALRR KLAHQRETGR DGPERHGPSS RGSLTPHPAA CDKDGQTD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRPF1
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRPF1
Gene Accession No. P55201
Gene Alias 4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140
Gene Symbols BRPF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRPF1. Recombinant protein control fragment (Product #RP-100748).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.