Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRPF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120519 Supplier Invitrogen™ Supplier No. PA5120519
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis. Immunogen sequence: MGVDFDVKTF CHNLRATKPP YECPVETCRK VYKSYSGIEY HLYHYDHDNP PPPQQTPLRK HKKKGRQSRP ANKQSPSPSE VSQSPGREVM SYAQAQRMVE VDLHGRVHRI SIFDNLDVVS EDEEAPEEAP ENGSNKENTE TPAATPKSGK HKNKEKRKDS NHHHHHNVSA STTPKLPEVV YRELEQDTPD APPRPTSYYR.

BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRPF1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BRPF1
Gene Accession No. B2RRD7, P55201
Gene Alias 4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140
Gene Symbols BRPF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human BRPF1 (NP_001003694).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 679713, 7862, 78783
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.