Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRPF1 Recombinant Rabbit Monoclonal Antibody (19H16L8)

Catalog No. 703489
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-348-9 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-348-9 Supplier Invitrogen™ Supplier No. 703489
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat, Bovine.

BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRPF1
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 19H16L8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene BRPF1
Gene Accession No. P55201
Gene Alias 4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140
Gene Symbols BRPF1
Host Species Rabbit
Immunogen Protein corresponding to human BRPF1 [aa1-aa100].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.