Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRPF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554605 Supplier Invitrogen™ Supplier No. PA554605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDNGINRLSL MAPDTPAGTP LSGVGRRTSV LFKKAKNGVK LQRSPDRVLE NGEDHGVAGS PASPASIEEE RHSRKRPRSR SCSESEGERS PQQEEETGMT NGFGKHTESG SDSECSLGLS GGLAFEACSG LTPPKRSRG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.

Scaffold subunit of various histone acetyltransferase (HAT) complexes, such as the MOZ/MORF and HBO1 complexes, which have a histone H3 acetyltransferase activity (PubMed:16387653, PubMed:26620551, PubMed:26677226). Plays a role in DNA replication initiation by directing KAT7/HBO1 specificity towards histone H3 'Lys-14' acetylation (H3K14ac), thereby facilitating the activation of replication origins (PubMed:26620551). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity (PubMed:16387653). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRPF3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRPF3
Gene Accession No. Q9ULD4
Gene Alias AI413466; AW493400; bromodomain and PHD finger containing 3; bromodomain and PHD finger containing, 3; bromodomain and PHD finger-containing protein 3; BRPF3; KIAA1286; mKIAA1286
Gene Symbols BRPF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRPF3. Recombinant protein control fragment (Product #RP-102480).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.