Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRPF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569705
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 92%.
Scaffold subunit of various histone acetyltransferase (HAT) complexes, such as the MOZ/MORF and HBO1 complexes, which have a histone H3 acetyltransferase activity (PubMed:16387653, PubMed:26620551, PubMed:26677226). Plays a role in DNA replication initiation by directing KAT7/HBO1 specificity towards histone H3 'Lys-14' acetylation (H3K14ac), thereby facilitating the activation of replication origins (PubMed:26620551). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity (PubMed:16387653). [UniProt].
Specifications
BRPF3 | |
Polyclonal | |
Unconjugated | |
BRPF3 | |
AI413466; AW493400; bromodomain and PHD finger containing 3; bromodomain and PHD finger containing, 3; bromodomain and PHD finger-containing protein 3; BRPF3; KIAA1286; mKIAA1286 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
268936 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B2KF05 | |
BRPF3 | |
synthetic peptide directed towards the middle region of mouse BRPF3. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction