Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRRN1Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991792
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1792 Supplier Rockland Immunochemicals Supplier No. 600401682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Reactivity occurs against human BRRN1 protein. However, BLAST analysis indicates 100% homology (18/18) for this protein from human, and chimpanzee. Partial cross reactivity is expected against mouse, dog, rat chicken, and from based on sequence homology of the amino terminal 13 amino acids of the immunogen sequence. The core 8 amino acids of the immunogen, A-G-P-D-H-W-R-F, show 100% identity with numerous proteins from a variety of organisms. If blots show multiple banding patterns in any particular lysate, it is suggested to confirm the specific reactivity against the BRRN1 protein using peptide competition. Reactivity with Brnn1 proteins from other sources is not known.

This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.

Specifications

Antigen BRRN1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene NCAPH
Gene Accession No. Q15003
Gene Alias A730011O11Rik; barren homolog 1; Barren homolog protein 1; BRRN; Brrn1; Caph; CAP-H; Chromosome-associated protein H; Condensin complex subunit 2; hCAP-H; KIAA0074; mCAP-H; mKIAA0074; NCAPH; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Gene Symbols NCAPH
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23397
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.