Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRSK1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298618JF549
Description
BRSK1 Polyclonal antibody specifically detects BRSK1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
BRSK1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
BR serine/threonine kinase 1, EC 2.7.11, EC 2.7.11.1, FLJ43009, KIAA1811BR serine/threonine-protein kinase 1, protein kinase SAD1A, SAD1, SAD1 kinase, Serine/threonine-protein kinase SAD-B | |
Produced in rabbits immunized with E. coli-derived Human BRSK1 fragment. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Signal Transduction | |
84446 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction