Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRSK1 Polyclonal Antibody
BRSK1 Polyclonal Antibody
Supplier: Thermo Scientific PA5101216
Description
BRSK1 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
BRSK1 | |
Polyclonal | |
Unconjugated | |
Brsk1 | |
BR serine/threonine kinase 1; BR serine/threonine-protein kinase 1; Brain-selective kinase 1; brain-specific serine/threonine-protein kinase 1; Brsk1; Gm1100; hSAD1; KIAA1811; protein kinase SAD1A; RGD1563268; SAD1; SAD1 homolog; SAD1 kinase; SAD1A; SADB; SAD-B; SadB kinase short isoform; serine/threonine kinase SADB; serine/threonine kinase SAD-B; serine/threonine-protein kinase BRSK1; Serine/threonine-protein kinase SAD-B; synapses of Amphids Defective homolog 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
381979, 84446 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5RJI5, Q8TDC3 | |
Brsk1 | |
A synthesized peptide derived from human BRSK1(Accession Q8TDC3), corresponding to amino acid residues Y352-V402. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction