Learn More
Invitrogen™ Human BRSK2, GST Tag Recombinant Protein

Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human BRSK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for BRSK2 is NP_003948.2.

Specifications
Specifications
Accession Number | Q8IWQ3 |
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 9024 |
Molecular Weight (g/mol) | 101 kDa |
Name | Human BRSK2, GST Tag |
Quantity | 100 μg |
Gene Alias | 4833424K13Rik; BR serine/threonine kinase 2; BR serine/threonine-protein kinase 2; brain serine/threonine kinase 2; brain-selective kinase 2; Brain-specific serine/threonine-protein kinase 2; BRSK2; C11orf7; HUSSY-12; Kiaa4256; PEN11B; protein kinase SAD1B; SAD1; SAD1B; Sada; SAD-A; serine/threonine-protein kinase 29; Serine/threonine-protein kinase BRSK2; serine/threonine-protein kinase SAD-A; STK29; testicular tissue protein Li 30 |
Common Name | STK29 |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.