Learn More
Invitrogen™ BRUCE Polyclonal Antibody

Description
The antibody detects endogenous levels of total BIRC6 protein.
Specifications
Specifications
| Antigen | BRUCE |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | BIRC6 |
| Gene Accession No. | Q9NR09 |
| Gene Alias | A430032G04Rik; A430040A19Rik; AA501170; APOLLON; baculoviral IAP repeat containing 6; baculoviral IAP repeat-containing 6; baculoviral IAP repeat-containing protein 6; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; accccbaculoviral IAP repeat-containing 6; BIR repeat-containing ubiquitin-conjugating enzyme; Birc6; BRUCE; D630005A10Rik; KIAA1289; mKIAA1289; RING-type E3 ubiquitin transferase BIRC6; Ubiquitin-conjugating BIR domain enzyme apollon; ubiquitin-conjugating BIR-domain enzyme apollon |
| Gene Symbols | BIRC6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.