Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brucella Abortus Antibody (BrG11), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073111AF700
Description
Brucella Abortus Monoclonal specifically detects Brucella Abortus in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Brucella Abortus | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
LPS of Brucella abortus and intact Brucella-cells. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA | |
BrG11 | |
Western Blot, ELISA | |
B.abortus | |
This is reacting with intact Brucella-cells. | |
0.2 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction