Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brucine dihydrate, 98%

Catalog No. p-7023150
Click to view available options
Quantity:
10 g
50 g

CAS: 5892-11-5 | C6H3FeK3O12 | 440.22 g/mol

Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

Solubility
Soluble in methanol. Partially soluble Miscible with water.

Notes
Incompatible with strong oxidizing agents, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5892-11-5
MDL Number MFCD00149384

Specifications

Melting Point 176°C to 178°C
Molecular Formula C6H3FeK3O12
Quantity 10 g
UN Number UN1570
Solubility Information Soluble in methanol. Partially soluble Miscible with water.
InChI Key YXTWCYYDGRBUDE-UHFFFAOYSA-K
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
IUPAC Name tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
Molecular Weight (g/mol) 440.22
PubChem CID 131675837
Formula Weight 430.50 (394.47 Anhydrous)
Percent Purity 98%
Chemical Name or Material Brucine dihydrate
Show More Show Less
Hazard Category H300+H330
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BRUCINE
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.