Learn More
Invitrogen™ Brugia malayi filaria Monoclonal Antibody (WES-7)

Specifications
Specifications
| Antigen | Brugia malayi filaria |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | WES-7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10mM sodium azide |
| Gene Alias | B. malayi; Filariasis; Philariasis |
| Host Species | Mouse |
| Immunogen | Native Brugia malayia filaria (roundworm). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.