Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRUNOL4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215622
Description
BRUNOL4 Polyclonal specifically detects BRUNOL4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BRUNOL4 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Bruno (Drosophila) -like 4, RNA binding protein, BRUNOL-4, BRUNOL4Bruno -like 4, RNA binding protein, bruno-like 4, RNA binding protein, bruno-like 4, RNA binding protein (Drosophila), Bruno-like protein 4, CELF-4, CUG-BP and ETR-3 like factor 4, CUG-BP- and ETR-3-like factor 4, CUGBP Elav-like family member 4, CUGBP, Elav-like family member 4, LYST-interacting protein LIP9, RNA-binding protein BRUNOL4, RNA-binding protein BRUNOL-4 | |
Rabbit | |
52 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (98%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.69mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9BZC1 | |
CELF4 | |
Recombinant protein encompassing a sequence within the center region of human BRUNOL4. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
56853 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction