Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRUNOL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113712 Supplier Invitrogen™ Supplier No. PA5113712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFSGVQQYTA MYPTAAITPI AHSVPQPPPL LQQQQREGPE GCNLFIYHLP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 90%; Pig: 100%; Rabbit: 79%; Rat: 90%; Yeast: 91%; Zebrafish: 100%.

Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternatively spliced transcript variants have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRUNOL5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CELF5
Gene Accession No. Q8N6W0
Gene Alias 4930565A21Rik; BRUNOL5; BRUNOL-5; bruno-like 5; bruno-like 5 RNA binding protein; bruno-like 5, RNA binding protein; Bruno-like protein 5; CELF5; CELF-5; CUG-BP and ETR-3 like factor 5; CUG-BP- and ETR-3-like factor 5; CUGBP Elav-like family member 5; CUGBP, Elav-like family member 5; RGD1565016; RNA-binding protein BRUNOL-5
Gene Symbols CELF5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human BRUNOL5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60680
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.