Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRUNOL6 (CELF6) Monoclonal Antibody (OTI2B8), OriGene
SDP

Catalog No. 502543881
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3881 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3881 Supplier OriGene Supplier No. CF811937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BRUNOL6 (CELF6) Monoclonal antibody specifically detects BRUNOL6 (CELF6) in Human samples. It is validated for Western Blot

Specifications

Antigen BRUNOL6 (CELF6)
Applications Western Blot
Classification Monoclonal
Clone OTI2B8
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CELF6
Gene Accession No. Q96J87
Gene Alias 6330569O16Rik; Bruno -like 6, RNA binding protein; BRUNOL6; bruno-like 6, RNA binding protein; bruno-like protein 6; CELF6; CELF-6; CUG-BP and ETR-3 like factor 6; CUG-BP- and ETR-3-like factor 6; CUGBP Elav-like family member 6; CUGBP, Elav-like family member 6; RNA-binding protein BRUNOL-6
Host Species Mouse
Immunogen Full length human recombinant protein of human CELF6 (NP_443072) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60677
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.