Learn More
PIP™ Brush Fire Helmet with ESS Goggle Mounts
PIP™ Brush Fire Helmet with ESS Goggle Mounts features a lightweight Aramid fibers composite resin shell for excellent strength and durability
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-046-414
|
PIP™
8696080 |
Red | N/A | N/A | N/A | ||||
19-046-415
|
PIP™
8696078 |
White | N/A | N/A | N/A | ||||
19-046-416
|
PIP™
8696081 |
Yellow | N/A | N/A | N/A | ||||
Description
- Composite resins offer higher temperature resistance (up to 500°F) without distorting or melting
- Shell weighs less than Thermoplastic equivalent models
- Polyurethane impact liner with leather sweatband provides impact and thermal protection
- Headband with removable padding for cleaning or replacement
- Ratcheting adjustable headband with merino padding for fit and comfort
- Leather reinforced mesh 6-point ribbon cradle
- ESS goggle mounts
- 3-Point Nomex™ chin strap with quick-release buckle keeps helmet secure
- High-gloss paint finish available in a variety of colors
- Meets ANSI/ISEA Z89.1-2009 requirements
- Certified to NFPA 1971, AS 4067
Specifications
Fire Protectant Helmets | |
One Size |
Warning - Prop 65 - Cancer and Reproductive Harm