Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRWD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558620
Description
Immunogen sequence: ILILDLEVNQ TVGVIAIERT GVPFLQVIPC FQRDGLFCLH ENGCITLRVR RSYNNIFTTS NEEPDPDPVQ ELTYDLRSQC D Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.
WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.
Specifications
BRWD2 | |
Polyclonal | |
Unconjugated | |
WDR11 | |
2900055P10Rik; AW489876; bromodomain and WD repeat domain containing 2; bromodomain and WD repeat-containing protein 2; BRWD2; DR11; HH14; Kiaa1351; mKIAA1351; sensitization to ricin complex subunit 1; SRI1; WD repeat domain 11; WD repeat domain 15; WD repeat-containing protein 11; WD repeat-containing protein 15; WDR11; WDR15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZH6 | |
WDR11 | |
Recombinant protein corresponding to Human BRWD2. Recombinant protein control fragment (Product #RP-97074). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction