Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRWD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588006 Supplier Invitrogen™ Supplier No. PA588006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YFQKFQLERV NLQEVKRSTY DHTRKCTDQL LLLGQTDRAV QLLLETSADN QHYYCDSLKA CLVTTVTSSG PSQSTIKLVA TNMIANGKLA EGVQLLCLID KAADACRYLQ TYGEWNRAAW LAKVRLNPEE CADVLRRWVD HLCSPQVNQK SKALLVLLSL GCFFSVAETL HSMRYFDRAA LFVEACLKYG AFEVTEDTEK LITAIYADYA RSLKNLGFKQ GAVLFASKAG AAGKDLLNEL ESPKEEPIEE; Positive Samples: DU145, HL-60, HepG2; Cellular Location: Cytoplasm, Membrane, Nucleus, Single-pass membrane protein.

WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRWD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WDR11
Gene Accession No. Q9BZH6
Gene Alias 2900055P10Rik; AW489876; bromodomain and WD repeat domain containing 2; bromodomain and WD repeat-containing protein 2; BRWD2; DR11; HH14; Kiaa1351; mKIAA1351; sensitization to ricin complex subunit 1; SRI1; WD repeat domain 11; WD repeat domain 15; WD repeat-containing protein 11; WD repeat-containing protein 15; WDR11; WDR15
Gene Symbols WDR11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 975-1224 of human WDR11 (NP_0605878).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55717
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.