Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRWD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101474
Description
Antibody detects endogenous levels of total BRWD3.
The protein encoded by this gene contains a bromodomain and several WD repeats. It is thought to have a chromatin-modifying function, and may thus play a role in transcription. Mutations in this gene cause mental retardation X-linked type 93, which is also referred to as mental retardation X-linked with macrocephaly. This gene is also associated with translocations in patients with B-cell chronic lymphocytic leukemia. [provided by RefSeq, May 2010].
Specifications
BRWD3 | |
Polyclonal | |
Unconjugated | |
Brwd3 | |
BRODL; bromo domain-containing protein disrupted in leukemia; bromodomain and WD repeat domain containing 3; bromodomain and WD repeat-containing protein 3; Brwd3; BRWD3-H; BRWD3-I; BRWD3-K; BRWD3-P; D030064D06Rik; Gm596; MRX93; novel WD repeat domain protein; RGD1559445 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
254065, 317213, 382236 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2AHJ4, Q6RI45 | |
Brwd3 | |
A synthesized peptide derived from human BRWD3(Accession Q6RI45), corresponding to amino acid residues S1763-Y1802. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction