Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRWD3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981687
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1687 Supplier Bioss Supplier No. BS11811R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene contains a bromodomain and several WD repeats. It is thought to have a chromatin-modifying function, and may thus play a role in transcription. Mutations in this gene cause mental retardation X-linked type 93, which is also referred to as mental retardation X-linked with macrocephaly. This gene is also associated with translocations in patients with B-cell chronic lymphocytic leukemia. [provided by RefSeq, May 2010].

Specifications

Antigen BRWD3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Brwd3
Gene Accession No. Q6RI45
Gene Alias Brodl; bromo domain-containing protein disrupted in leukemia; bromodomain and WD repeat domain containing 3; Bromodomain and WD repeat-containing protein 3; Brwd3; BRWD3-H; BRWD3-I; BRWD3-K; BRWD3-P; D030064D06Rik; Gm596; MRX93; novel WD repeat domain protein; RGD1559445
Gene Symbols Brwd3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BRWD3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254065
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.