Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Bryostatin 1
GREENER_CHOICESDP

Catalog No. 238310U Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-831-0U 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-831-0U Supplier R&D Systems™ Supplier No. 2383/10U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase C activator

Protein kinase C (PKC) activator that binds with high affinity (Ki = 1.35 nM). Initially activates and subsequently induces downregulation of PKC isozymes. Sensitizes tumor cells to cytotoxic effects of anticancer agents. Restores hippocampal synapses and spatial learning and memory in an in vivo model of fragile X syndrome.

Chemical Identifiers

CAS 83314-01-6
Molecular Formula C47H68O17

Specifications

Quantity 10 μg
Product Line Tocris
Formula Weight 905.04
Purity >98%
Molecular Weight (g/mol) 905.044
Chemical Name or Material Bryostatin 1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.