Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BS 181 dihydrochloride
GREENER_CHOICE

Catalog No. 560810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
560810 10 mg
560850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 560810 Supplier Tocris Bioscience™ Supplier No. 5608/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective cdk7 inhibitor

BS 181 dihydrochloride is a selective cdk7 inhibitor (IC50 = 21 nM). Displays >40-fold selectivity for cdk7 over other cdks. Promotes cell cycle arrest and apoptosis in a range of cancer cells in vitro and inhibits MCF-7 tumor xenograft growth in mice.

Specifications

Chemical Name or Material N5-(6-Aminohexyl)-3-(1-methylethyl)-N7-(phenylmethyl)pyrazolo[1,5-a]pyrimidine-5,7-diamine dihydrochloride
CAS 1883548-83-1
Quantity 10 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C22H32N6.2HCl
Formula Weight 453.45
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.